Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Aprotinin, MP Biomedicals™

Used largely as an inhibitor of trypsin. Aprotinin, MP Biomedicals is a competitive serine protease inhibitor that forms stable complexes with and blocks the active sites of enzyme. This binding is reversible, and most aprotinin-protease complexes will di

$878.00 - $3410.00

Specifications

Form Solution
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP218056310
View Documents
MP Biomedicals™
0218056310
10 mL
Each for $878.00
Add to cart
 
MP218056350
View Documents
MP Biomedicals™
0218056350
50 mL
Each for $3,410.00
Add to cart
 
Description

Description

Structurally, Aprotinin is a monomeric globular protein derived from bovine lung that consists of 58 amino acids, arranged in a single polypeptide chain with three cross-linking disulfide bridges.

Specifications

Specifications

Solution
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit