Learn More
Ansell™ APS Splash Suit Boots
Yellow PVC or black nitrile safety boots provide good chemical resistance to a wide range of industrial chemicals. Boots are fixed to splash suits with an ergonomically designed ring attachment, for simplified boot exchange. Available in sizes 8-13.
Supplier: Ansell™ 072203470
This item is not returnable.
View return policy
Description
- PVC or Nitrile Safety Boot providing good chemical resistance
- Features anti-slip sole and anti-static properties
- Steel toecap, steel midsole and oil resistance sole
- Soft, synthetic inner lining provides comfort in use for the wearer
- PVC boot is standard used on TRELLCHEM™ gas tight and splash suits
- Nitrile rubber safety boots with European approval as Firemen boots (not on suits certified to NFPA 1991:2016)
Specifications
Yellow | |
Splash suits | |
PVC | |
13 |