Learn More
Aat BioquestSupplier Diversity Partner APTS [8-Aminopyrene-1,3,6-trisulfonic acid, trisodium salt] *CAS 196504-57-1* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 624
This water-soluble reagent has strong green fluorescence with great Stokes shift. It is widely used for detecting carbohydrates and labeling glycoproteins in electrophoresis.
Catalog No. 50-195-2940