Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AQP1 Monoclonal Antibody (OTI1F3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Supplier: OriGene TA502358
Description
Aquaporin 1 is a 28kD integral membrane protein which was originally identified in red blood cells and renal proximal tubules. AQP1 is also expressed by the choroid plexus and various other tissues. It forms a water-specific channel that provides the plasma membranes of red cells and kidney proximal tubules with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient.Specifications
AQP1 | |
Monoclonal | |
0.7 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P29972 | |
AQP1 | |
Full length human recombit protein of human AQP1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI1F3 | |
Unconjugated | |
AQP1 | |
AQP-CHIP, CHIP28, CO | |
Mouse | |
Affinity Chromatography | |
RUO | |
358 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction