Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AQP11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578807
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue, HELA whole cell.
Aqp11 encodes aquaporins (AQPs) that are integral membrane proteins that facilitate the transport of water across biological membranes along an osmotic gradient. There have been 13 AQP isoforms (AQP0-AQP12) identified in humans and rodents to date. Aquaporins facilitate the transport of water and small neutral solutes across cell membranes.
Specifications
AQP11 | |
Polyclonal | |
Unconjugated | |
AQP11 | |
1700015P13Rik; AI648923; AQP11; AQP-11; AQPX1; aquaporin 11; aquaporin-11; CG12251 gene product; PSEC0027; sjds | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
282679, 286758, 66333 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
Q8BHH1, Q8CHM1, Q8NBQ7 | |
AQP11 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human AQP11 (35-70aa ARQQLHRPVAHAFVLEFLATFQLCCCTHELQLLSEQ). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction