Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AQPEP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549459

Catalog No. PA549459

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Metalloprotease which may be important for placentation by regulating biological activity of key peptides at the embryo- maternal interface. On synthetic substrates it shows a marked preference for Leu-4-methylcoumaryl-7-amide (Leu-MCA) over Met- MCA, Arg-LCA and Lys-LCA. Cleaves the N-terminal amino acid of several peptides such as angiotensin-3, kisspeptin-10 and endokinin C.
Specifications

Specifications

AQPEP
Polyclonal
PBS with 0.09% sodium azide
Q6Q4G3
Aminopeptidase Q, AP-Q, 3411-, CHL2 antigen, Laeverin, AQPEP, LVRN
Rabbit
IgG
80 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
206338
Western Blot
Unconjugated
LVRN
Liquid
LVRN
KLH conjugated synthetic peptide between 844-877 amino acids from human AQPEP.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit