Thermo Scientific™ AquaBath™ Water Baths

Aquabath Water Baths provide precise temperature control and are available with an analog or digital operating system, in a variety of sizes from 2 to 28L.

Manufacturer: Thermo Scientific™ 18102AQ

 View more versions of this product

Catalog No. 15-475-35Q

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Hertz 50/60Hz
    Length (English) Exterior 16.4 in.
    Length (Metric) Exterior 42cm
    Shipping Weight (Metric) 10.1kg
    Temperature Range (Metric) Ambient to 100°C
    Temperature Uniformity ±0.24° at 37°C
    Width (English) Exterior 24.2 in.
    Width (English) Interior 19.8 in.
    Width (Metric) Exterior 62cm
    Capacity (English) 5.3 gal.
    Capacity (Metric) 20L
    Control Digital Control
    Length (English) Interior 11.8 in.
    Length (Metric) Interior 30cm
    Certifications/Compliance CSA and CE certified
    Dimensions (L x W x H) Exterior 16.4 x 24.2 x 8.9 in. (42 x 62 x 23cm)
    Voltage 120V
    Cabinet Material Stainless-steel shell with enamel coating
    Chamber Material Stainless steel
    Depth (Metric) Interior 15cm
    Height (Metric) Exterior 23cm
    Shipping Weight (English) 22.3 lb.
    Width (Metric) Interior 50cm
    Capacity 5.3 gal., 20L
    Electrical Requirements 120V 50/60Hz
    Depth (English) Interior 6 in.
    Height (English) Exterior 8.9 in.

    Thermo Scientific™ Aquabath Water Baths provide precise temperature control and are available with an analog or digital operating system. The Aquabath is available in a variety of sizes from 2 to 28L, including a shallow form to meet specific application needs and budgets. Bath features dual thermostats and an independent high-limit thermostat for overtemperature protection.

    • Hinged acrylic cover lifts to a 90° stay-open position or can be removed completely
    • “Fins” on the hinged clear acrylic gable cover (included) protect hands from hot vapors
    • Exterior remains cool to the touch even after extended use to prevent accidental burns
    • Temperature-resistant plastic rim fits over the seamless stainless-steel reservoir for a leak-free seal
    • Stainless-steel exterior resists corrosion and has a tough, easy-to-clean enamel coating
    • Rounded, seamless stainless-steel reservoir and diffuser tray resist rust and contamination for simple maintenance
    • Movable temperature controllers on the 20 and 28L digital models can be used on the bath long or short side to best utilize bench space
    • Heating elements will not burn out if the bath accidentally runs dry
    • Drain pump is included with 10, 20, 28 and 5/10L models

    Digital Operating System

    • Field-calibratable
    • PID control provides ±0.24°C uniformity, ±0.5°C stability and ±0.1°C control resolution at 37°C
    • Digital temperature set and readout selectable display with 0.1° resolution

    Analog Operating System

    • Easy-to-use analog control and hydraulic thermostat—ideal for fixed-setpoint applications
    • Temperature uniformity ±0.2°C at 37°C
    • Diffuser tray, thermometer clip and nonhazardous liquid, partial immersion (76mm) thermometer (-10° to +110°C) included (230V models do not include a thermometer)

    Ordering Information:

    Stainless-steel covers available separately.

    Compatible with:

    • Cover, for 2L baths, stainless steel (19000-11Q)
    • Cover, for 2L shallow and 5L baths, stainless steel (19000-13Q)
    • Cover, for 20L and 28L baths, stainless steel (19000-17Q)
    • Cover, for 10L baths, stainless steel (19000-15Q)

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit