Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AquaBath™ Water Baths

Aquabath Water Baths provide precise temperature control and are available with an analog or digital operating system, in a variety of sizes from 2 to 28L.

Manufacturer:  Thermo Scientific™ 18002AQ

 View more versions of this product

Catalog No. 22-299-732Q

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications

Description

Thermo Scientific™ Aquabath Water Baths provide precise temperature control and are available with an analog or digital operating system. The Aquabath is available in a variety of sizes from 2 to 28L, including a shallow form to meet specific application needs and budgets. Bath features dual thermostats and an independent high-limit thermostat for overtemperature protection.

  • Hinged acrylic cover lifts to a 90° stay-open position or can be removed completely
  • “Fins” on the hinged clear acrylic gable cover (included) protect hands from hot vapors
  • Exterior remains cool to the touch even after extended use to prevent accidental burns
  • Temperature-resistant plastic rim fits over the seamless stainless-steel reservoir for a leak-free seal
  • Stainless-steel exterior resists corrosion and has a tough, easy-to-clean enamel coating
  • Rounded, seamless stainless-steel reservoir and diffuser tray resist rust and contamination for simple maintenance
  • Movable temperature controllers on the 20 and 28L digital models can be used on the bath long or short side to best utilize bench space
  • Heating elements will not burn out if the bath accidentally runs dry
  • Drain pump is included with 10, 20, 28 and 5/10L models

Digital Operating System

  • Field-calibratable
  • PID control provides ±0.24°C uniformity, ±0.5°C stability and ±0.1°C control resolution at 37°C
  • Digital temperature set and readout selectable display with 0.1° resolution

Analog Operating System

  • Easy-to-use analog control and hydraulic thermostat—ideal for fixed-setpoint applications
  • Temperature uniformity ±0.2°C at 37°C
  • Diffuser tray, thermometer clip and nonhazardous liquid, partial immersion (76mm) thermometer (-10° to +110°C) included (230V models do not include a thermometer)

Ordering Information:

Stainless-steel covers available separately.

Compatible with:

  • Cover, for 2L baths, stainless steel (19000-11Q)
  • Cover, for 2L shallow and 5L baths, stainless steel (19000-13Q)
  • Cover, for 20L and 28L baths, stainless steel (19000-17Q)
  • Cover, for 10L baths, stainless steel (19000-15Q)
Description & Specifications

Specifications

Ambient to 100°C
Stainless-steel shell with enamel coating
6 in.
7.5 in.
50/60Hz
27cm
4.5kg
14.5 in.
37cm
5L
6 in.
CSA and CE certified
120V
Stainless steel
15cm
19cm
10.5 in.
10 lb.
±0.24° at 37°C
11.8 in.
30cm
Digital
15cm
10.5 x 14.5 x 7.5 in. (27 x 37 x 19cm)
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit