Learn More
Karl Fischer Aqualine™ Complete 1, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Single-component reagent; Pyridine free
This item is not returnable.
View return policy
Description
- Single-component volumetric reagent for high water content analysis
- Water equivalent: 1.0mg H2O/mL
- Unlimited water capacity
- Perform titer determination before use
- Pyridine free
Specifications
Colorless | |
1 L | |
Karl Fischer Reagent | |
1.2mg of water/mL min. | |
Liquid | |
81734 |
Aqualine™ Complete 1 | |
Pyridine free | |
111-90-0, 288-32-4, 7446-09-5, 7553-56-2 | |
Amber glass bottle | |
Solution |
Safety and Handling
DANGER!
Emergency Overview
Irritating to skin. Irritating to eyes. Possible risk of harm to the unborn child.
NFPA
Health: 1
Signal Word: DANGER!