Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Aqualine™ Matrix K, for Karl Fischer Titration By Volumetry, Fisher Chemical™

 View more versions of this product

Catalog No. AL2300R-1


Description

Description

  • Matrix to be used in conjunction with Complete 5K (AL2250R-1) for high water content analysis of aldehydes and ketones
Specifications

Specifications

107-21-1,111-76-2,5683-33-0
Colorless
Karl Fischer Reagent
Liquid
Pass test
Aqualine™ Matrix K
81734
Amber glass bottle
1L
> 1.0
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Causes burns by all exposure routes. May cause central nervous system effects. Possible risk of harm to the unborn child. May form explosive peroxides. Use personal protective equipment. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:1
Instability:1

DOTInformation : DOT Class 6.1, : Poison

SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit