Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Aqualine™ Solvent, for Karl Fischer Titration By Volumetry, Fisher Chemical™

Solvent for two-component system; Pyridine free

 View more versions of this product

Catalog No. AL-21001


Order in Bulk

Request bulk or custom formats

Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Solvent for two-component system; Pyridine free

  • Used in conjunction with Aqualine™ titrant for two component volumetric Karl Fischer analysis
Specifications

Specifications

Aqualine™ Solvent
1L
Karl Fischer Reagent
Amber glass bottle
> 1.0
1992
Colorless
Pyridine free
288-32-4, 7446-09-5
Pass Test
81734
Liquid
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

DOTInformation : DOT Class 3, : Flammable Liquid

SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit