Learn More
Aquaporin 1/AQP1 Antibody (OTI2D10), FITC, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270199F
Catalog No. NBP270199F
Add to Cart
Description
Description
Aquaporin 1/AQP1 Monoclonal specifically detects Aquaporin 1/AQP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Aquaporin 1/AQP1
Monoclonal
FITC
PBS with 0.05% Sodium Azide
AQP1
Full length recombinant protein of human Aquaporin 1/AQP1 (NP_932766) produced in HEK293T cells.
0.1 mL
358.0
Store at 4°C in the dark.
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI2D10
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
28-kDa, AQP-1, AQP-CHIP, aquaporin 1 (channel-forming integral protein, 28kDa), aquaporin 1 (Colton blood group), Aquaporin-CHIP, CHIP2828kDa, CO blood group), CO, MGC26324
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG1
Product Suggestions
Product Suggestions