Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin 1/AQP1 Antibody (OTI2D10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270199H
Description
Aquaporin 1/AQP1 Monoclonal specifically detects Aquaporin 1/AQP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Aquaporin 1/AQP1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
AQP1 | |
Full length recombinant protein of human Aquaporin 1/AQP1 (NP_932766) produced in HEK293T cells. | |
0.1 mL | |
358.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2D10 | |
Western Blot, Immunohistochemistry | |
28-kDa, AQP-1, AQP-CHIP, aquaporin 1 (channel-forming integral protein, 28kDa), aquaporin 1 (Colton blood group), Aquaporin-CHIP, CHIP2828kDa, CO blood group), CO, MGC26324 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction