Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin 1 Rabbit anti-Bovine, Human, Mouse, Ovine, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc AQP1101AP

Description
Aquaporin 1 Polyclonal antibody specifically detects Aquaporin 1 in Bovine, Human, Mouse, Ovine, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
Aquaporin 1 | |
Polyclonal | |
Unconjugated | |
Aqp1 | |
AQP 1; AQP CHIP; Aqp1; AQP-1; AQP-CHIP; aquaporin 1; Aquaporin 1 (aquaporin channel forming integral protein 28 (CHIP)); aquaporin 1 (channel-forming integral protein, 28kDa, CO blood group); aquaporin 1 (Colton blood group); aquaporin 1, Colton blood group antigen; Aquaporin CHIP; Aquaporin1; aquaporin-1; Aquaporin-CHIP; channel-like integral membrane protein, 28-kDa; CHIP 28; CHIP28; CO; Colton blood group; Delayed early response protein 2; DER2; growth factor-induced delayed early response protein; membrane channel; MGC26324; urine water channel; Water channel protein for red blood cells and kidney proximal tubule | |
Synthetic peptide taken within amino acid region 215-265 on human Aquaporin 1 protein. | |
200 μL | |
Primary | |
Bovine, Human, Mouse, Ovine, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P29972, P29975, P47865, P56401, Q02013 | |
Rabbit | |
Affinity chromatography | |
RUO | |
11826, 25240, 282653, 358, 442999 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction