Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin-4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP197928
Description
Aquaporin-4 Polyclonal specifically detects Aquaporin-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Aquaporin-4 | |
Polyclonal | |
Unconjugated | |
PBS, 50% Glycerol with 0.09% Sodium Azide | |
aquaporin 4, MGC22454 | |
Rabbit | |
Affinity Purified | |
RUO | |
361 | |
Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1:1000-1:4000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:400 | |
NP_036957.1 | |
AQP4 | |
Produced against the C-terminal peptide (Sequence N-CTKGSYMEVEDNRSQVETED) of rat aquaporin 4 | |
0.1 mL | |
Primary | |
Detects approx 35kDa. May detect larger glycosylated bands approx 50kDa. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction