Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin-4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP312902

Catalog No. NB176589


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

Aquaporin-4 Polyclonal antibody specifically detects Aquaporin-4 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry - Free Floating
Specifications

Specifications

Aquaporin-4
Polyclonal
PBS, 1% BSA, 20% Glycerol (pH7)
Rabbit
Antigen Affinity-purified
RUO
Primary
Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating)
Unconjugated
aquaporin 4, MGC22454
Recombinant protein encompassing a sequence within the C-terminus region of mouse Aquaporin-4. The exact sequence is proprietary.
100 μg
Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
361
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.