Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin 9 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551285

Catalog No. PA551285


Add to cart

Description

Description

The antibody detects endogenous levels of total Aquaporin 9 protein.

The aquaporins are a family of water-selective membrane channels. The protein encoded by this gene allows passage of a wide variety of noncharged solutes. It stimulates urea transport and osmotic water permeability; there are contradicting reports about its role in providing glycerol permeability. The encoded protein may also play a role in specialized leukocyte functions such as immunological response and bactericidal activity.
Specifications

Specifications

Aquaporin 9
Polyclonal
Unconjugated
AQP9
Liquid
AQP9
Synthetic peptide of human AQP9
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
O43315
SSC1, AQP-9, HsT17287
Rabbit
IgG
100 μL
-20°C
Primary
366
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit