Learn More
R&D Systems™ AQUApure Tetra-Ub Chains (K33-linked), Carrier-Free Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Supplier:  R&D Systems™ UC103025
Catalog No. UC103025
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
The AQUApure Tetra-Ub Chains (K33-linked) Protein is derived from E. coli. The AQUApure Tetra-Ub Chains (K33-linked) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
P0CG47
7314
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
UBB
Unconjugated
E. coli-derived
Bioactivity
Theoretical M.W.: 34kDa
E. coli-derived human Tetra-Ubiquitin protein
Tetra-Ubiquitin
Ubiquitin chains vary in length, linkage, and function. K33-linked Tetra-Ubiquitin Chains (Ub4) are ideal for investigating Ubiquitin-binding proteins and as substrates for Ubiquitin-specific isopeptidases. Reaction conditions will need to be optimized for each specific application. IMPORTANT: Heating this product in SDS-PAGE buffer or terminating reactions containing this product with heated SDS-PAGE buffer could lead to unexpected, high apparent molecular weight banding or smearing on gels that is not representative of product purity. For optimal results, we recommend incubation in SDS-PAGE buffer + DTT at <40°C for 20 minutes prior to gel electrophoresis.
Human
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. Ub-AQUA analysis: K33: 95.08% K48: 2.69% K63: 0.90% K11: 0.71% All other linkages ≤ 0.38%
Product Suggestions
Product Suggestions