Learn More
Encon™ Aquarion™ Fluid Preserved Cartridges
Sealed ready to use cartridges that ensure safe, pure, contaminant-free flushing fluid. Encon™ Aquarion™ Fluid Preserved Cartridge is self-evacuating and stores the fluid while maintaining the required flow without any external force.
Manufacturer:  Encon™ AQ120
Catalog No. 19-811-303
Add to cart
Description
Description
  • The sealed ready to use cartridges ensure safe, pure, contaminant-free flushing fluid for 2 years
  • The self-evacuating cartridges store the fluid and help maintain a required flow throughout the entire 15-minute flush without any external force
  • The actuation is a simple, one-step process - just pull the tray down in an easy motion and the fluid begins to flow in less than a second
  • Sealed Fluid Cartridges
  • Preserved Saline Solution
Specifications
Specifications
3.7 gal per cartridge
Aquarion™ Self-Contained Portable Eyewash
Emergency eyewash saline replacement cartridges
70 lb.
14 L per cartridge
Saline Solution: Purified Water, Sodium Chloride, Boric Acid, Sodium Tetraborate, Chlorhexidine Digluconate
70 lb.