Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AquaStar Solvent MC, MilliporeSigma
Use with Composite 1, 2, 2K, 5, or 5K reagents
Specifications
Chemical Name or Material | AquaStar™ Solvent MC |
---|---|
Name Note | Reagent for Volumetric Karl Fischer Titrations |
CAS | 67-66-3 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
MAX1699MC1
|
MilliporeSigma
AX1699MC1 |
1L | Glass Bottle |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
Description
For titration of oils and fatsSpecifications
AquaStar™ Solvent MC | |
Reagent for Volumetric Karl Fischer Titrations | |
67-66-3 |
Safety and Handling
DOTInformation : DOT Class 3, : Flammable Liquid, Toxic
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title