Learn More
AR Mouse anti-Human, Clone: AR441 + DHTR/882, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB14823
Catalog No. 89-034-906
Add to cart
Description
Description
Mouse monoclonal antibody raised against human AR.
  • High specificity
  • High reactivity
Specifications
Specifications
AR
AR441 + DHTR/882
Mouse monoclonal antibody raised against human AR.
In 1 mM PBS.
Liquid
AR
A synthetic peptide corresponding to amino acids 302-318 of human AR (AR441) and recombinant protein corresponding to full length human AR (DHTR/882).
Protein A/G purification
RUO
Primary
367
Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
P10275
AIS|DHTR|HUMARA|KD|NR3C4|SBMA|SMAX1|TFM
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)