Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Arabidopsis thaliana ZAR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117660
Description
This antibody has been tested in direct-ELISA.
This maternal effect gene is oocyte-specific and encodes a protein that is thought to function in the initiation of embryogenesis. A similar protein in mouse is required for female fertility. [provided by RefSeq, Jul 2013].
Specifications
Arabidopsis thaliana ZAR1 | |
Polyclonal | |
Unconjugated | |
oocyte-specific maternal effect factor; Z3CXXC6; ZAR1; zinc finger, 3CxxC-type 6; zygote arrest 1; zygote arrest protein 1 | |
Recombinant Arabidopsis thaliana Receptor protein kinase-like protein ZAR1 protein (167-278aa). | |
100 μg | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
Western Blot | |
2.66 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Plant | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction