Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arachidonyl serotonin, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Dual fatty acid amide hydrolase (FAAH) inhibitor/TRPV1 antagonist (IC50 values are 5.6 μM and 37 - 40 nM for FAAH and TRPV1 respectively). Inactive at cPLA2, CB1 or 5-HT receptors. Displays strong analgesic activity against both acute and chronic peripheral pain.
Specifications
Quantity | 10 mg |
Formula Weight | 462.67 |
Percent Purity | >98% |
Chemical Name or Material | Arachidonyl serotonin |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction