Learn More
ARAP1 Mouse anti-Human, DyLight™ 550, Clone: ARAP1-2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11068801R
Catalog No. NB11068801S
Add to cart
Description
Description
ARAP1 Monoclonal specifically detects ARAP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
ARAP1
ARAP1-2
Western Blot, Immunocytochemistry/Immunofluorescence
Purified, Conjugated
ARAP1
His6-tagged protein corresponding to amino acids 1190-1450 of human ARAP1
Protein A purified
Cell Cycle and Replication
Primary
116985.0
Human
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 550
50 mM sodium borate with 0.05% sodium azide
arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1, centaurin, delta 2, Centaurin-delta-2, CENTD2ARF-GAP, RHO-GAP, ankyrin repeat, and pleckstrin homology domains-containingprotein 1, cnt-d2, KIAA0782centaurin-delta-2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
The ARAP1-2 reacts with C-terminal part of ARAP1, a 160 kDa adaptor protein.