Learn More
ARAP3 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 225781AP
Catalog No. 50-173-3893
Add to cart
Description
Description
Ubiquitin is a conserved 76 amino acid polypeptide and can affect proteasomal degradation of the protein it is bound to, or mediate interactions with other proteins related to post-translational modifications. The degradation of cellular regulatory proteins by the Uubiquitin pathway is important as it controls the cellular growth and proliferation. Ubiquitin-dependent proteolysis occurs after a covalent attachment of the peptide to a lysine residue of a protein, which involves three enzymatic reactions: E1, E2 and E3. The first reaction involves ubiquitin-activating enzyme. The third reaction uses enzyme ubiquitin ligase (E3) to transfer the activated ubiquitin from E2 to a lysine residue on a protein, or directly transfers the ubiquitin from E2 to the substrate.
Specifications
Specifications
ARAP3
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8WWN8
ARAP3, Centaurin delta 3, CENTD3, Cnt d3, DRAG1
Rabbit
IgG
150 μL
-20°C
Primary
64411
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ARAP3
Liquid
ARAP3
ARAP3 Fusion Protein Ag17436
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human