Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arbutin, 99.5%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
50 g
1 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
497-76-7
497-76-7
Packaging Fiber Drum
Loss on Drying 0.005
Quantity 25 kg
CAS Max % 1
Percent Purity 0.995
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.