Learn More
National Safety Apparel™ Arc H2O FR Hi-Vis Orange Rain Jacket - Type R Class 3 Tall
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Arc Rating: 19 cal/cm² (Fluorescent Yellow) | 24 cal/cm² (Fluorescent Orange)
- CAT 2
- Fabric/Material: PU Coated FR Cotton
- Waterproof welded seams with reinforced stress points to provide durability
- FR fabric will not melt, drip or ignite
- FR polyurethane is durable and flexible that will not become brittle as it ages
- Contoured collar with tuck away attached hood
- Double storm flap with reflective trim
- Adjustable cuffs
- Back ventilation system with FR mesh for increased airflow
- Covered D-ring opening for opening access
- Two reinforced front pockets with closure for convenient secure storage
- 2 in. silver reflective trim
- FR hook and loop tab at wrist cuff for improved fit
- Adjustable elastic suspender system for fit and comfort
- Covered zipper and snap closures
- Made in the USA
Specifications
Size | 3X-Large |
Material | Polyurethane-coated FR Cotton |
Color | Flourescent Orange |
Closure Type | Snap, Zipper |
Certifications/Compliance | Arc Rating: 19 cal/cm² (Fluorescent Yellow); 24 cal/cm² (Fluorescent Orange), CAT 2, NFPA 70 E, ANSI/ISEA 107 Type R Class 3, CSA Z462, ASTM F2733, ASTM F1891, OSHA 1910.269 |
Seam Type | Welded |