Learn More
National Safety Apparel™ Arc H2O FR Hi-Vis Yellow Rain Jacket - Type R Class 2
Arc H2O FR HI-Vis rain jacket type R class 2
Manufacturer:  National Safety Apparel™ R30RL053X
 View more versions of this product
Catalog No. 17-001-2057
Add to cart
Description
Description
  • Arc Rating: 19 cal/cm² (Fluorescent Yellow) | 24 cal/cm² (Fluorescent Orange)
  • CAT 2
  • Fabric/Material: PU Coated FR Cotton
  • Waterproof welded seams with reinforced stress points to provide durability
  • Batwing pattern creating freedom of movement
  • Contoured collar with tuck away
  • Covered zipper and snap closures
  • FR hook and loop adjustable tab closures
  • Back ventilation system with FR mesh for increased airflow
  • Covered D-Ring opening access
  • Two reinforced front pockets with FR hook and loop closure
  • 2 in. silver reflective trim
  • Adjustable cuffs
  • Front storm flap with reflective trim
  • FR polyurethane is durable and flexible that will not become brittle as it ages
  • FR fabric will not melt, drip, or ignite
  • Made in the USA
Specifications
Specifications
Arc Rating: 19 cal/cm², CAT 2, NFPA 70 E, ANSI/ISEA 107 Type R Class 2, CSA Z462, ASTM F2733, ASTM F1891, OSHA 1910.269
Flourescent Yellow
Welded
None
Zipper, Snap
Polyurethane-coated FR Cotton
3X-Large