Learn More
National Safety Apparel™ Arc H2O FR Rainwear Kit in Fluorescent Orange - Type R Class 3
Includes type R class 3 FR jacket, Class E FR bib overall, and black mesh gear bag
Supplier: National Safety Apparel™ KITRQC3LG
Description
- Arc Rating: 19 cal/cm² (Fluorescent Yellow) | 24 cal/cm² (Fluorescent Orange)
- CAT 2
- Fabric/Material: PU Coated FR Cotton
- 2 in. FR silver reflective trim
- FR polyurethane (PU) moisture barrier
- Lightweight and stretchable comfort
- FR polyurethane is durable and flexible that will not become brittle as it ages
- Waterproof welded seams with reinforced stress points to provide durability
- Back ventilation system with FR mesh for increased airflow
- Contoured collar with tuck away attached hood
- Covered D-Ring opening access
- Covered zipper and snap closure
- FR hook and loop adjustable leg closures
- Adjustable cuffs and shoulder straps
- Heavy duty quick release suspender system
- FR fabric will not melt, drip, or ignite
- Made in the USA
Specifications
CAT 2, NFPA 70 E, ANSI/ISEA 107 Type R Class 3, CSA Z462, ASTM F2733, ASTM F1891, OSHA 1910.269 | |
Fluorescent Orange | |
Adjustable | |
Polyurethane-coated FR Cotton | |
Welded |
19 cal/cm², 24 cal/cm² | |
Large | |
Open | |
Zipper, Snap |