Learn More
National Safety Apparel™ ArcGuard™ Revolite 40 Cal Long Coat Kit With Gloves (08)
Flash kit with short coat and bib overall
Manufacturer:  National Safety Apparel™ KIT4LCLT404X08
 View more versions of this product
Catalog No. 17-001-1572
Add to cart
Description
Description
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 11.2 oz. RevoLite™
  • 50% lighter than our standard 40 cal kits
  • FR hook and loop closure with drawstring waist
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Made in the USA
Specifications
Specifications
Gloves
FR Hook and Loop
Olive Green
Stitched
Adjustable
Arc Rating: 40 cal/cm², CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Open
RevoLite™
4X-Large