Learn More
National Safety Apparel™ ArcGuard™ Revolite 40 Cal Long Coat Kit With Gloves (08)
Flash kit with short coat and bib overall
$1963.47 - $2112.47
Specifications
Closure Type FR Hook and Loop
Color Olive Green
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-1542
View Documents
National Safety Apparel™
KIT4LCLT40SM08
Small
Each for $1,963.47
Add to cart
17-001-1547
View Documents
National Safety Apparel™
KIT4LCLT40MD08
Medium
Each for $1,963.47
Add to cart
17-001-1552
View Documents
National Safety Apparel™
KIT4LCLT40LG08
Large
Each for $1,963.47
Add to cart
17-001-1557
View Documents
National Safety Apparel™
KIT4LCLT40XL08
X-Large
Each for $1,963.47
Add to cart
17-001-1562
View Documents
National Safety Apparel™
KIT4LCLT402X08
2X-Large
Each for $2,013.48
Add to cart
17-001-1567
View Documents
National Safety Apparel™
KIT4LCLT403X08
3X-Large
Each for $2,013.48
Add to cart
17-001-1572
View Documents
National Safety Apparel™
KIT4LCLT404X08
4X-Large
Each for $2,062.46
Add to cart
17-001-1577
View Documents
National Safety Apparel™
KIT4LCLT405X08
5X-Large
Each for $2,112.47
Add to cart
Description
Description
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 11.2 oz. RevoLite™
  • 50% lighter than our standard 40 cal kits
  • FR hook and loop closure with drawstring waist
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Made in the USA
Specifications
Specifications
FR Hook and Loop
Olive Green