Learn More
National Safety Apparel™ ArcGuard™ Revolite 40 Cal Long Coat Kit With Gloves (09)
Flash kit with short coat and bib overall
$1963.47 - $2112.47
Specifications
Seam Type Stitched
Material RevoLite™
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-1543
View Documents
National Safety Apparel™
KIT4LCLT40SM09
Small
Each for $1,963.47
Add to cart
17-001-1548
View Documents
National Safety Apparel™
KIT4LCLT40MD09
Medium
Each for $1,963.47
Add to cart
17-001-1553
View Documents
National Safety Apparel™
KIT4LCLT40LG09
Large
Each for $1,963.47
Add to cart
17-001-1558
View Documents
National Safety Apparel™
KIT4LCLT40XL09
X-Large
Each for $1,963.47
Add to cart
17-001-1563
View Documents
National Safety Apparel™
KIT4LCLT402X09
2X-Large
Each for $2,013.48
Add to cart
17-001-1568
View Documents
National Safety Apparel™
KIT4LCLT403X09
3X-Large
Each for $2,013.48
Add to cart
17-001-1573
View Documents
National Safety Apparel™
KIT4LCLT404X09
4X-Large
Each for $2,062.46
Add to cart
17-001-1578
View Documents
National Safety Apparel™
KIT4LCLT405X09
5X-Large
Each for $2,112.47
Add to cart
Description
Description
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 11.2 oz. RevoLite™
  • 50% lighter than our standard 40 cal kits
  • FR hook and loop closure with drawstring waist
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Made in the USA
Specifications
Specifications
Stitched
RevoLite™