Learn More
National Safety Apparel™ ArcGuard™ Revolite 40 Cal Long Coat Kit With Gloves (10)
Flash kit with short coat and bib overall
$1963.47 - $2112.47
Specifications
Seam Type Stitched
Material RevoLite™
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-1544
View Documents
National Safety Apparel™
KIT4LCLT40SM10
Small
Each for $1,963.47
Add to cart
17-001-1549
View Documents
National Safety Apparel™
KIT4LCLT40MD10
Medium
Each for $1,963.47
Add to cart
17-001-1554
View Documents
National Safety Apparel™
KIT4LCLT40LG10
Large
Each for $1,963.47
Add to cart
17-001-1559
View Documents
National Safety Apparel™
KIT4LCLT40XL10
X-Large
Each for $1,963.47
Add to cart
17-001-1564
View Documents
National Safety Apparel™
KIT4LCLT402X10
2X-Large
Each for $2,013.48
Add to cart
17-001-1569
View Documents
National Safety Apparel™
KIT4LCLT403X10
3X-Large
Each for $2,013.48
Add to cart
17-001-1574
View Documents
National Safety Apparel™
KIT4LCLT404X10
4X-Large
Each for $2,062.46
Add to cart
17-001-1579
View Documents
National Safety Apparel™
KIT4LCLT405X10
5X-Large
Each for $2,112.47
Add to cart
Description
Description
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 11.2 oz. RevoLite™
  • 50% lighter than our standard 40 cal kits
  • FR hook and loop closure with drawstring waist
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Made in the USA
Specifications
Specifications
Stitched
RevoLite™