Learn More
Epredia™ ArcosSL™ Accessories
Epredia™ ArcosSL™ slide management system accessories can be used to provide pathologists with flexible networking solutions for labs with multiple users.
Manufacturer:  Epredia™ A85310037
 View more versions of this product
Catalog No. A85310037
Add to cart
Description
Description
Epredia™ ArcosSL™ slide management system is designed to enhance sample traceability throughout the sorting, archiving, retrieval and re-archiving processes. The additional PDA and/or Arcos app license options work in conjunction with the system for situations where multiple users are required.  The ArcosSL accessories can be configured to meet the connectivity needs of laboratories to minimize errors and increase productivity.
Data Security and Network Flexibility
  • ArcosSL features redundant protection. Its database backup is automatically updated on the ArcosSL PC and Flash storage within the scanner. The unit can be configured to automatically archive your database backup to your choice of secure storage – hard drive or external network.
  • Laboratories have the ability to control individual user database access and modification permissions.
  • Benefit from flexible network-based or LIS-based connectivity options to meet your laboratory’s configuration:
    • Network-based Connectivity:
      • Data back up
      • Multiple PDAs
      • Multiple scanners
      • Arcos app (for multiple users (PCs))
    • LIS-based Connectivity:
      • Uni-directional connection
      • Bi-directional connection
  • Additional PDAs are helpful for labs with multiple users. The PDAs work in conjunction with the system by sending the picking list from the main Epredia™ ArcosSL™ station to the additional PDAs.
  • Multi-PDAs enable users to send the picking list from ArcosSL to any PDA location, eliminating the need to bring the PDA to the main lab. The synchronization is done via the network.  This is ideal for labs where multiple users are required.
  • The Arcos app includes a bi-directional communication feature, linking a specific workstation to the lab and enabling data access to remote PCs. This supports multi-user configuration where each app license can run reports, create picking lists and dispatch them to other users.
Peace of Mind
  • With proper use of the PDA, ArcosSL is designed to reduce the risk of retrieving the wrong slide, misplacing a slide, filing slides incorrectly, and/or losing a slide.
  • Rapid retrieval of the correct slide can reduce impact to patient safety.
  • ArcosSL features are designed to reduce the risk of errors endemic to hand-sorting processes.
Specifications
Specifications
ArcosSL™ Safety Bars 6/Pk
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Epredia™ Arcos™ app must be operated with Arcos software 1.3.