Learn More
Agilent Technologies ARCTIC EXPRESSDE3 RIL COMPCELL
Supplier: Agilent Technologies 230193
Enhanced protein folding and solubility of expressed proteins. Dramatically improves expression for AT-rich genomes when codon bias is a problem. Encodes T7 RNA polymerase under control of the lacUV5 promoter.