Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARD1A Monoclonal Antibody (06), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Scientific MA543042

Encompass

 View more versions of this product

Catalog No. PIMA543042


Add to cart
This item is not returnable. View return policy

Description

Description

Specificity: Human ARD1

ARD1A Monoclonal antibody specifically detects ARD1A in Human samples. It is validated for ELISA
Specifications

Specifications

ARD1A
Monoclonal
1 mg/mL
PBS with no preservative
P41227
NAA10
Recombinant Human ARD1 protein.
100 μL
Primary
Human
Antibody
IgG1
ELISA
06
Unconjugated
NAA10
2310039H09Rik; alpha-N-acetyltransferase 1A; ARD1; ARD1 homolog A, N-acetyltransferase; Ard1a; ARD1P; DXS707; hARD1; MCOPS1; N(alpha)-acetyltransferase 10, NatA catalytic subunit; N(alpha)-acetyltransferase 10, NatA catalytic subunitNalpha acetyltransferase 10; Naa10; N-acetyltransferase ARD1 homolog; N-acetyltransferase ARD1, human homolog of; Nalpha acetyltransferase 10; N-alpha-acetyltransferase 10; N-alpha-acetyltransferase 10, NatA catalytic subunit; natA catalytic subunit Naa10; NATD; N-terminal acetyltransferase complex ARD1 subunit homolog A; OGDNS; RGD1565315; Te2
Mouse
Protein A
RUO
8260
-20° C, Avoid Freeze/Thaw Cycles
Liquid
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit