Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARD1A Monoclonal Antibody (06), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA543486
This item is not returnable.
View return policy
Description
Specificity: Human ARD1
ARD1A Monoclonal antibody specifically detects ARD1A in Human samples. It is validated for ELISA.Specifications
ARD1A | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P41227 | |
NAA10 | |
Recombinant Human ARD1 protein. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
06 | |
Unconjugated | |
NAA10 | |
2310039H09Rik; alpha-N-acetyltransferase 1A; ARD1; ARD1 homolog A, N-acetyltransferase; Ard1a; ARD1P; DXS707; hARD1; MCOPS1; N(alpha)-acetyltransferase 10, NatA catalytic subunit; N(alpha)-acetyltransferase 10, NatA catalytic subunitNalpha acetyltransferase 10; Naa10; N-acetyltransferase ARD1 homolog; N-acetyltransferase ARD1, human homolog of; Nalpha acetyltransferase 10; N-alpha-acetyltransferase 10; N-alpha-acetyltransferase 10, NatA catalytic subunit; natA catalytic subunit Naa10; NATD; N-terminal acetyltransferase complex ARD1 subunit homolog A; OGDNS; RGD1565315; Te2 | |
Mouse | |
Protein A | |
RUO | |
8260 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction