Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARD1A Mouse anti-Human, Monkey, Mouse, Clone:3G3E9, Abnova™

Mouse monoclonal antibody raised against recombinant human ARD1A.

Manufacturer:  Abnova Corporation MAB17498

Catalog No. 89-333-927


Add to cart

Description

Description

N-alpha-acetylation is one of the most common protein modifications that occurs during protein synthesis and involves the transfer of an acetyl group from acetyl-coenzyme A to the protein alpha-amino group. ARD1A, together with NATH (NARG1; MIM 608000), is part of a major N-alpha-acetyltransferase complex responsible for alpha-acetylation of proteins and peptides (Sanchez-Puig and Fersht, 2006 [PubMed 16823041]).[supplied by OMIM]
Specifications

Specifications

ARD1A
3G3E9
ARD1 homolog A, N-acetyltransferase (S. cerevisiae)
In PBS (0.05% sodium azide)
Liquid
Mouse
IgG1
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Monkey, Mouse
ELISA, Immunocytochemistry, Western Blot
Unconjugated
ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry (1:200-1:1000)
Flow Cytometry
Immunohistochemistry
The optimal working dilution should be determined by the end user.
ARD1A
ARD1, DXS707, MGC71248, TE2
Recombinant protein corresponding to amino acid 111-235 of human ARD1A from E. coli.
100μg
Antibody
8260
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit