Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF1 Mouse anti-Human, Allophycocyanin/Cy7, Clone: 1B3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP197935APCCY7

Catalog No. N197935APC7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ARF1 Monoclonal antibody specifically detects ARF1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

ARF1
1B3
Flow Cytometry
Conjugated, Purified
ARF1
Recombinant human ARF1 (1-181aa) purified from E. coli
Protein G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Anti-human ARF1 mAb, clone 1B3, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human ARF1 protein.
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
ADP-ribosylation factor 1
Mouse
IgG1 Kappa
0.1 mL
Primary
375.0
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit