Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARF1 Rabbit anti-Human, Clone: SD2002, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267739
Description
ARF1 Monoclonal specifically detects ARF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ARF1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100 | |
ADP-ribosylation factor 1 | |
Rabbit | |
Protein A purified | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
SD2002 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
ARF1 | |
Synthetic peptide within Human ARF1aa 61-110/ 181. (SwissProt: P84077 Human; SwissProt: P84078 Mouse; SwissProt: P84079 Rat) | |
100 μL | |
375 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction