Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARF3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30448220UL
Description
ARF3 Polyclonal specifically detects ARF3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ARF3 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Unconjugated | |
ADP-ribosylation factor 3, small GTP binding protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-181 of human ARF3 (NP_001650.1). | |
20 μL | |
Signal Transduction | |
377 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction