Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen ARF3
Applications Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095961
View Documents
Novus Biologicals
NBP304482100UL
100 μg Each for $320.71
Add to cart
 
NB095962
View Documents
Novus Biologicals
NBP30448220UL
20 μg Each for $138.37
Add to cart
 
Description

Description

ARF3 Polyclonal antibody specifically detects ARF3 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

ARF3
Unconjugated
Rabbit
Signal Transduction
PBS with 50% glycerol, pH7.3.
377
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence
Purified
RUO
Human
ADP-ribosylation factor 3, small GTP binding protein
Recombinant fusion protein containing a sequence corresponding to amino acids 1-181 of human ARF3 (NP_001650.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit