Learn More
Cell Biolabs IncSupplier Diversity Partner ARF6 ACTIVATION KT 20 ASSY  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA4076
The Arf6 Activation Assay Kit provides a non-radioactive assay format to pull down the active form of Ras with colored agarose beads, which allow for a visual check, followed by Western blot with a Arf6 antibody. 20 assays/kit.
Catalog No. 50-673-609