Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARFGAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$496.07

Specifications

Antigen ARFGAP2
Concentration 0.34 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-173-1003
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
165191AP150UL
150 μL
Each of 1 for $496.07
Add to Cart
 
Description

Description

ARFGAP2 gene ontology annotations related to this gene include GTPase activator activity; metal ion binding.
Specifications

Specifications

ARFGAP2
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
Q3MID3, Q8N6H7, Q99K28
362162, 77038, 84364
ARFGAP2 Fusion Protein Ag9740
Primary
-20°C
ARFGAP2
0.34 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
ARF GAP 2, ARFGAP2, IRZ, Nbla10535, Zfp289, Zinc finger protein 289, ZNF289
Arfgap2
IgG
Antigen Affinity Chromatography
Antibody
SDS
Documents

Documents

Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit