Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARG2 Antibody (OTI3G5), FITC, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270205F

Catalog No. NBP270205F


Add to Cart

Description

Description

ARG2 Monoclonal specifically detects ARG2 in Human, Rat, Canine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ARG2
Monoclonal
FITC
PBS with 0.05% Sodium Azide
ARG2
Full length recombinant protein of human ARG2 (NP_001163) produced in HEK293T cells.
0.1 mL
Primary
Human, Rat, Canine
IgG1
Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI3G5
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
arginase 2, arginase, type II, arginase-2, mitochondrial, EC 3.5.3, EC 3.5.3.1, kidney arginase, Kidney-type arginase, L-arginine amidinohydrolase, L-arginine ureahydrolase, nonhepatic arginase, Non-hepatic arginase, Type II arginase
Mouse
Affinity Purified
Lipid and Metabolism
384.0
Store at 4°C in the dark.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit