Learn More
Abnova™ Arg8-Vasopressin ELISA Kit
For the Quantitative detection of Human Arg8-Vasopressin.
Supplier: Abnova™ KA4582
Description
- Arg8-Vasopressin ELISA Kit is a competitive immunoassay for the quantitative determination of Vasopressin in samples.
Specifications
Human | |
1 Kit | |
Human |
Kit | |
Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing. |