Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARHGEF10L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555307
Description
Immunogen sequence: ALEHSEEDGS IYEMADDPDV WVRSRPCARD AHRKEICSVA IISGGQGYRN FGSALGSSGR QAPCGETDST LLIWQVPL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
ARHGEF10L (Rho Guanine Nucleotide Exchange Factor 10 Like) is a Protein Coding gene. Among its related pathways areRET signalingandG protein signaling_RhoB regulation pathway. Gene Ontology (GO) annotations related to this gene includeRho guanyl nucleotide exchange factor activity. An important paralog of this gene isARHGEF10.
Specifications
ARHGEF10L | |
Polyclonal | |
Unconjugated | |
ARHGEF10L | |
argal; Arhgef10l; GrinchGEF; KIAA1626; Rho guanine nucleotide exchange factor (GEF) 10-like; Rho guanine nucleotide exchange factor 10 like; rho guanine nucleotide exchange factor 10-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55160 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCE6 | |
ARHGEF10L | |
Recombinant protein corresponding to Human ARGAL. Recombinant protein control fragment (Product #RP-94017). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction