Learn More
Arginase 1/ARG1/liver Arginase Antibody (OTI4E6), DyLight 405, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270204V
Catalog No. NBP270204V
Add to Cart
Description
Description
Arginase 1/ARG1/liver Arginase Monoclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
Specifications
Arginase 1/ARG1/liver Arginase
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
ARG1
Full length recombinant protein of human Arginase 1/ARG1/liver Arginase (NP_000036) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Flow Cytometry, Immunohistochemistry
OTI4E6
Western Blot, Flow Cytometry, Immunohistochemistry
arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase
Mouse
Affinity Purified
Cancer, Cellular Markers, Chromatin Research, Lipid and Metabolism, Myeloid derived Suppressor Cell
383.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions