Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arginase 1/ARG1/liver Arginase Rabbit anti-Human, Mouse, Rat, PE, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132731PE

Catalog No. NBP132731PE


Add to cart

Description

Description

Arginase 1/ARG1/liver Arginase Polyclonal antibody specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

Arginase 1/ARG1/liver Arginase
PE
PBS with 0.05% Sodium Azide
arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human, Mouse, Rat
Flow Cytometry
Flow Cytometry
Affinity Purified, Conjugated
ARG1
Arginase 1/ARG1/liver Arginase Antibody made from full length human Arginase 1 Recombinant protein.
Immunogen affinity purified
Cancer, Cellular Markers, Chromatin Research, Lipid and Metabolism, Myeloid derived Suppressor Cell
Primary
383.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit