Learn More
Arginase 1 Monoclonal Antibody (A1exF5), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$169.00
Specifications
Antigen | Arginase 1 |
---|---|
Clone | A1exF5 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
25-369-780
|
eBioscience
25369780 |
25 μg |
Each for $169.00
|
|
|||||
25-369-782
|
eBioscience
25369782 |
100 μg |
Each for $169.00
|
|
|||||
Description
Description: The monoclonal antibody A1exF5 recognizes both human and mouse Arginase 1, a cytosolic enzyme (Arg1). This A1exF5 clone is compatible with both, the standard intracellular protocols, and the Foxp3/Transcription Factor Staining Buffer Set. Applications Reported: This A1exF5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This A1exF5 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product No. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No.
Arginase 1 Monoclonal antibody specifically detects Arginase 1 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Arginase 1 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AI; A-I; AI type I arginase; AI256583; Arg1; Arg-1; Arginase; arginase 1; arginase 1 liver; arginase 1, liver; arginase I; arginase, liver; Arginase1; arginase-1; HGNC:663; Liver Arginase; Liver-type arginase; PGIF; similar to arginase, type I; Type 1 Arginase; type I arginase | |
ARG1 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
A1exF5 | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Human, Mouse | |
P05089, Q61176 | |
11846, 383 | |
E.coli-derived recombinant mouse Arginase 1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ARG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.